Palladium(II)kloridi (59% Pd)

Toimittaja: Thermo Fisher Scientific
Vaara

195200010 195200050 195200250
ACRO195200010EA 242 EUR
ACRO195200010 ACRO195200050 ACRO195200250
Palladium(II)kloridi (59% Pd)
Palladium(II)kloridi
Kaava: PdCl₂
Tiheys: 4 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003558
CAS-numero: 7647-10-1
EINECS: 231-596-2
UN: 3260
ADR: 8,III
Merck Index: 13,07058

Order Now

Testitulosten erittely

Appearance Red-brown Powder
Assay 59 to 60 % (Pd)
Calcium (Ca) ≤30 ppm
Copper (Cu) ≤15 ppm
Iron (Fe) ≤15 ppm
Lead (Pb) ≤15 ppm
Magnesium (Mg) ≤15 ppm
Silicon (Si) ≤30 ppm
Silver (Ag) ≤30 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR