Rubidiumkloridi 99.99% (trace metals basis)

Toimittaja: Thermo Fisher Scientific

Synonyms: Rubidiumchloride

193920100
ACRO193920100EA 302 EUR
ACRO193920100
Rubidiumkloridi 99.99% (trace metals basis)
Rubidiumkloridi
Kaava: RbCl
Kiehumispiste: 1390 °C (760 mmHg)
Sulamispiste: 718 °C
Tiheys: 2,8 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011187
CAS-numero: 7791-11-9

Order Now

Testitulosten erittely

Appearance White Powder and/or chunks
Titration Argentometric 28.9 to 30.1 % (Cl)
Total trace metal impurities ≤100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR