Osmiumtetraoksidi 99.9+% (trace metals basis)

Toimittaja: Thermo Fisher Scientific
Vaara

191180010 191181000 191182500 191185000 191180050
ACRO191180010EA 342 EUR
ACRO191180010 ACRO191181000 ACRO191182500 ACRO191185000 ACRO191180050
Osmiumtetraoksidi 99.9+% (trace metals basis)
Osmiumtetraoksidi
Kaava: O₄Os
Kiehumispiste: 130 °C (760 mmHg)
Sulamispiste: 40…42 °C
Tiheys: 1,04 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011150
CAS-numero: 20816-12-0
EINECS: 244-058-7
UN: 2471
ADR: 6.1,I
Merck Index: 13,06962

Order Now

Testitulosten erittely

Appearance Yellow Crystals
Melting point 40 to 42°C
Heavy metals ≤0.005 %
Solubility Passes test
Purity ≥99.90 % (Metal purity)
Non-volatile matter ≤0.2 %
Osmium (Os) 74.2 to 75.4 %
Ruthenium (Ru) ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR