Trypan blue

Toimittaja: Thermo Fisher Scientific
Vaara

189350250 189351000 189355000
ACRO189350250EA 51.9 EUR
ACRO189350250 ACRO189351000 ACRO189355000
Trypan blue
Trypan blue
Kaava: C₃₄H₂₄N₆Na₄O₁₄S₄
Sulamispiste: >280 °C (haj. lämpötila)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003969
CAS-numero: 72-57-1
EINECS: 200-786-7

Order Now

Testitulosten erittely

Appearance Dark blue to brown-black Powder
Infrared spectrum Conforms
UV spectrum Complies
Dye content ≥60 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR