3,4,5-Trimethoxyphenol 98.5+%

Toimittaja: Thermo Fisher Scientific
Varoitus

Synonyms: Antiarol

189140050 189140250 189141000
ACRO189140050EA 92.8 EUR
ACRO189140050 ACRO189140250 ACRO189141000
3,4,5-Trimethoxyphenol 98.5+%
3,4,5-Trimethoxyphenol
Kaava: C₉H₁₂O₄
Kiehumispiste: 283 °C (1013 hPa)
Sulamispiste: 146…149 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00008389
CAS-numero: 642-71-7
EINECS: 211-387-2

Order Now

Testitulosten erittely

Appearance White to yellow to beige to brown Powder or crystals or needles
Infrared spectrum Conforms
Melting point 143 to 149 °C
GC ≥98.50 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR