α,α,α-Trifluori-p-tolueenihappo 98%

Toimittaja: Thermo Fisher Scientific
Varoitus

188460010 188460050
ACRO188460010EA 25.9 EUR
ACRO188460010 ACRO188460050
α,α,α-Trifluori-p-tolueenihappo 98%
α,α,α-Trifluori-p-tolueenihappo
Kaava: C₈H₅F₃O₂
Sulamispiste: 219…222 °C
MDL numero: MFCD00002562
CAS-numero: 455-24-3
EINECS: 207-242-8

Order Now

Testitulosten erittely

Appearance White to light grey Crystalline powder
Infrared spectrum Conforms
Melting point 219 to 222 °C
GC ≥97.5 %
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR