2,3,5,6-Tetrafluorifenoli 98%

Toimittaja: Thermo Fisher Scientific
Vaara

188390050 188390250
ACRO188390050EA 56.5 EUR
ACRO188390050 ACRO188390250
2,3,5,6-Tetrafluorifenoli 98%
2,3,5,6-Tetrafluorifenoli
Kaava: C₆H₂F₄O
Kiehumispiste: 140 °C (1013 hPa)
Sulamispiste: 33…38 °C
Leimahduspiste: 79 °C
MDL numero: MFCD00002157
CAS-numero: 769-39-1
EINECS: 212-209-6
UN: 2923
ADR: 8,III

Order Now

Testitulosten erittely

Appearance White Low melting crystalline solid
Infrared spectrum Conforms
Melting point 33 to 38 °C
GC ≥97.5 %
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR