Polyvinyylialkoholi, M.W. ∼ 146,000-186,000 92-94%, hydrolysed

Toimittaja: Thermo Fisher Scientific

183160010 183160025 183161000
ACRO183160010EA 185 EUR
ACRO183160010 ACRO183160025 ACRO183161000
Polyvinyylialkoholi, M.W. ∼ 146,000-186,000 92-94%, hydrolysed
Polyvinyylialkoholi
Kaava: [-CH₂CH(OH)-]n
Sulamispiste: 230…240 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00081922
CAS-numero: 9002-89-5

Order Now

Testitulosten erittely

Appearance White to cream Granular powder
Infrared spectrum Conforms
Viscosity 53 to 63 cP (4 % in water)
Degree of hydrolysis 92.0 to 94.0 mol %
Ash ≤0.6 % (as Na2O)
pH 4.5 to 6.5 (4 % in water)
Volatile matter ≤5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR