Formamidi 99.5+%, extra pure

Toimittaja: Thermo Fisher Scientific
Vaara

181090010 181090025 181090250
ACRO181090010EA 60.4 EUR
ACRO181090010 ACRO181090025 ACRO181090250
Formamidi 99.5+%, extra pure
Formamidi
Kaava: HCONH₂
Kiehumispiste: 210 °C (760 mmHg)
Sulamispiste: 2…3 °C
Tiheys: 1,133 g/cm³
Leimahduspiste: 175 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00007941
CAS-numero: 75-12-7
EINECS: 200-842-0
Merck Index: 13,04261
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f381691a96baf5054ca8291e1fa9e9603b687e31&vid=e54dda1a985d7a4f9485c2cc5233498087e43f88&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4460 to 1.4480 (20°C, 589 nm)
Water ≤0.1 % (Coulometric)
GC ≥99.50 %
Residue after evaporation ≤0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR