Pyreeni 98%

Toimittaja: Thermo Fisher Scientific
Varoitus

180830250 180831000 180835000
ACRO180830250EA 42.5 EUR
ACRO180830250 ACRO180831000 ACRO180835000
Pyreeni 98%
Pyreeni
Kaava: C₁₆H₁₀
MW: 202.26 g/mol
Kiehumispiste: 393 °C (760 mmHg)
Sulamispiste: 148…152 °C
Leimahduspiste: 210 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00004136
CAS-numero: 129-00-0
EINECS: 204-927-3
UN: 3077
ADR: 9 III
Merck Index: 12,08147
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4b03eb6b87b60315a282988993f066fd2d3af129&vid=d3f1ffa103d650d2e9f49139db04842dfa6ca0ff&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Light yellow to yellow to yellow-green Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 147 to 152 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR