3,3-Dimethylbutylamine 95%

Toimittaja: Thermo Fisher Scientific
Vaara

179380010 179380050
ACRO179380010EA 114 EUR
ACRO179380010 ACRO179380050
3,3-Dimethylbutylamine 95%
3,3-Dimethylbutylamine

Appearance: Clear colorless to yellow Liquid

Kaava: C₆H₁₅N
Kiehumispiste: 114…116 °C (760 mmHg)
Tiheys: 0,752 g/cm³
Leimahduspiste: 6 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00008201
CAS-numero: 15673-00-4
UN: 3286
ADR: 3,II

Order Now

Testitulosten erittely

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4120 to 1.4160 (20°C, 589 nm)
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR