STYRENE, M.W. ∼ 260,000

Toimittaja: Thermo Fisher Scientific

178890010 178890250 178895000 178891000
ACRO178890010EA 400 EUR
ACRO178890010 ACRO178890250 ACRO178895000 ACRO178891000
STYRENE, M.W. ∼ 260,000
Poly(styrene)

CAS No.: 9003-53-6

Kaava: C₈H₈
MW: 125000 - 250000 g/mol
Sulamispiste: 93 °C
Tiheys: 1.05 g/cm³
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00084450
CAS-numero: 9003-53-6

Order Now

Testitulosten erittely

Appearance Clear Pellets
Infrared spectrum Conforms
Loss on drying ≤2 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR