Metanoli 99+%, extra pure

Toimittaja: Thermo Fisher Scientific
Vaara

Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol

177150010 177150025 177150250 177150050
ACRO177150010EA 39.1 EUR
ACRO177150010 ACRO177150025 ACRO177150250 ACRO177150050
Metanoli 99+%, extra pure
Metanoli
Kaava: CH₃OH
Kiehumispiste: 64,6 °C (1013 hPa)
Sulamispiste: –98 °C
Tiheys: 0,791 g/cm³
Leimahduspiste: 12 °C (sulj. astia)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00004595
CAS-numero: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Clear liquid
Water ≤0.2 % (K.F.)
GC ≥99.0 %
Color scale ≤10 APHA
Residue after evaporation ≤0.005 %
Calcium (Ca) ≤0.005 %
Copper (Cu) ≤0.001 ù
Iron (Fe) ≤0.002 %
Lead (Pb) ≤0.005 %
Magnesium (Mg) ≤0.005 %
Manganese (Mn) ≤0.0005 %
Mercury (Hg) ≤0.000001 %
Nickel (Ni) ≤0.0005 %
Potassium (K) ≤0.005 %
Sodium (Na) ≤0.005 %
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR