Puuvillasiemenöljy, pure

Toimittaja: Thermo Fisher Scientific

177115000 177110025
ACRO177115000EA 56.9 EUR
ACRO177115000 ACRO177110025
Puuvillasiemenöljy, pure
Puuvillasiemenöljy
Tiheys: 0,91 g/cm³
Leimahduspiste: >112 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00130872
CAS-numero: 8001-29-4
EINECS: 232-280-7

Order Now

Testitulosten erittely

Appearance Yellow Oily liquid
Infrared spectrum Conforms
Refractive index 1.4720 to 1.4730 (20°C, 589 nm)
Additional info linolenic and higher: ≤1.5%
Acid value ≤0.5 mg KOH/g
Iodine value 95 to 115 g I2/100 g
Saponification value 185 to 198 mg KOH/g
Unsaponifiable matter ≤1.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR