3-Nitrofenoli 99%

Toimittaja: Thermo Fisher Scientific
Vaara

172300100 172300500 172302500
ACRO172300100EA 53.9 EUR
ACRO172300100 ACRO172300500 ACRO172302500
3-Nitrofenoli 99%
3-Nitrofenoli
Kaava: C₆H₅NO₃
Kiehumispiste: 194 °C (70 mmHg)
Sulamispiste: 94…98 °C
Tiheys: 1,49 g/cm³
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00007240
CAS-numero: 554-84-7
EINECS: 209-079-8
UN: 1663
ADR: 6.1,III
Merck Index: 12,06682

Order Now

Testitulosten erittely

Appearance Yellow to brown Crystalline powder
Infrared spectrum Conforms
Melting point 94 to 98 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR