Methyl bromoacetate 99%

Toimittaja: Thermo Fisher Scientific
Vaara

169550025 169550250 169551000 169555000
ACRO169550025EA 336 EUR
ACRO169550025 ACRO169550250 ACRO169551000 ACRO169555000
Methyl bromoacetate 99%
Methyl bromoacetate
Kaava: BrCH₂COOCH₃
Kiehumispiste: 145 °C (1013 hPa)
Sulamispiste: –50 °C
Tiheys: 1,61 g/cm³
Leimahduspiste: 62 °C
Storage Temperature: Jääkaappi
MDL numero: MFCD00000189
CAS-numero: 96-32-2
EINECS: 202-499-2
UN: 2643
ADR: 6.1,II

Order Now

Testitulosten erittely

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4570 to 1.4590 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR