2-Etyyliaminoetanoli 98%

Toimittaja: Thermo Fisher Scientific
Vaara

168730010 168732500
ACRO168730010EA 68.4 EUR
ACRO168730010 ACRO168732500
2-Etyyliaminoetanoli 98%
2-Etyyliaminoetanoli
Kaava: C₄H₁₁NO
Kiehumispiste: 167 °C (1013 hPa)
Sulamispiste: –6 °C
Tiheys: 0,91 g/cm³
Leimahduspiste: 78 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00002841
CAS-numero: 110-73-6
EINECS: 203-797-5
UN: 2735
ADR: 8,III

Order Now

Testitulosten erittely

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4390 to 1.4430 (20°C, 589 nm)
Water ≤0.5 %
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR