2-Fluori-5-nitroaniliini 98%

Toimittaja: Thermo Fisher Scientific
Varoitus

168390250 168391000 168390050
ACRO168390250EA 69.3 EUR
ACRO168390250 ACRO168391000 ACRO168390050
2-Fluori-5-nitroaniliini 98%
2-Fluori-5-nitroaniliini
Kaava: C₆H₅FN₂O₂
Sulamispiste: 99…103 °C
Leimahduspiste: 91 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00007652
CAS-numero: 369-36-8
EINECS: 206-720-3
UN: 2811
ADR: 6.1,III

Order Now

Testitulosten erittely

Appearance Ochre to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 99 to 103 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR