2,4,6-Tris(2-pyridyl)-1,3,5-triazine 99%

Toimittaja: Thermo Fisher Scientific
Varoitus

168070050 168070010 168070250 168071000
ACRO168070050EA 192 EUR
ACRO168070050 ACRO168070010 ACRO168070250 ACRO168071000
2,4,6-Tris(2-pyridyl)-1,3,5-triazine 99%
2,4,6-Tris(2-pyridyl)-1,3,5-triazine
Kaava: C₁₈H₁₂N₆
Sulamispiste: 248…252 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00006045
CAS-numero: 3682-35-7
EINECS: 222-965-9

Order Now

Testitulosten erittely

Appearance White to cream to yellow Powder
Infrared spectrum Conforms
Melting point 248 to 252 °C
Titration with HClO4 98.5 to 101.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR