3-Piperidinemethanol 98%

Toimittaja: Thermo Fisher Scientific
Vaara

Synonyms: 3-(Hydroxymethyl)piperidine

168040050 168040250 168041000
ACRO168040050EA 28.6 EUR
ACRO168040050 ACRO168040250 ACRO168041000
3-Piperidinemethanol 98%
3-Piperidinemethanol
Kaava: C₆H₁₃NO
Kiehumispiste: 236 °C (760 mmHg)
Sulamispiste: 58…62 °C
Tiheys: 1,02 g/cm³
Leimahduspiste: >112 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00005997
CAS-numero: 4606-65-9
UN: 3263
ADR: 8,II

Order Now

Testitulosten erittely

Appearance White to yellow Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 56 to 63 °C
GC ≥97.5 %
Iron (Fe) ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR