1-Butanoli 99+% for spectroscopy

Toimittaja: Thermo Fisher Scientific
Vaara

167691000 167695000 167690025
ACRO167691000EA 54.5 EUR
ACRO167691000 ACRO167695000 ACRO167690025
1-Butanoli 99+% for spectroscopy
1-Butanoli
Kaava: H₃C(CH₂)₂CH₂OH
Kiehumispiste: 117,6 °C (760 mmHg)
Sulamispiste: –89 °C
Tiheys: 0,81 g/cm³
Leimahduspiste: 35 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00002964
CAS-numero: 71-36-3
EINECS: 200-751-6
UN: 1120
ADR: 3,III
REACH: 01-2119484630-38
Merck Index: 13,01539
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9395eeb6150a74222da52460f53cedeccd93442&vid=fd8352a556914c5be7213c576355a88e130d1a7b&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Clear liquid
Water ≤0.05 % (Coulometric)
GC ≥99.0 %
UV 1 cm cell vs HPLC-grade water
Color scale ≤10 APHA
Residue after evaporation ≤0.0005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR