Akryyliamidi 98.5%, extra_pure

Toimittaja: Thermo Fisher Scientific
Vaara

Synonyms: Prop-2-eeniamidi

164830025 164835000 164830100
ACRO164830025EA 150 EUR
ACRO164830025 ACRO164835000 ACRO164830100
Akryyliamidi 98.5%, extra_pure
Akryyliamidi
Kaava: C₃H₅NO
MW: 71.08 g/mol
Kiehumispiste: 125 °C (25 mmHg)
Sulamispiste: 82…86 °C
Tiheys: 1.122 g/cm³
Leimahduspiste: 138 °C
Storage Temperature: Jääkaappi
MDL numero: MFCD00008032
CAS-numero: 79-06-1
EINECS: 201-173-7
UN: 2074
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7687b7f17a8cdf31c559a33fd5d4021a2e50a40a&vid=fecebe0c34b037788df119c11bfc9637d78cda0d&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 82 to 86 °C
Water ≤2.0 %
GC ≥98.0 %
Color of solution (10 % in water) Clear and colorless


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR