4-Phenylpiperidine 98%

Toimittaja: Thermo Fisher Scientific
Varoitus

164770010 164770050
ACRO164770010EA 54.1 EUR
ACRO164770010 ACRO164770050
4-Phenylpiperidine 98%
4-Phenylpiperidine
Kaava: C₁₁H₁₅N
MW: 161.25 g/mol
Kiehumispiste: 255…257 °C (727 mmHg)
Sulamispiste: 57…63 °C
Tiheys: 0.967
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00006002
CAS-numero: 771-99-3
EINECS: 212-243-1

Order Now

Testitulosten erittely

Appearance Off-white to light brown Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 59 to 64 °C
GC ≥97.5 %

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR