7-Amino-1,3-naphthalenedisulphonic acid tech.

Toimittaja: Thermo Fisher Scientific
Vaara

163961000 163965000 163960025
ACRO163961000EA 51.9 EUR
ACRO163961000 ACRO163965000 ACRO163960025
7-Amino-1,3-naphthalenedisulphonic acid tech.
7-Amino-1,3-naphthalenedisulphonic acid

Appearance: Off-white to green-beige to light brown Powder or needles

Kaava: C₁₀H₁₂N₂O₆S₂
Sulamispiste: >300 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003991
CAS-numero: 86-65-7
UN: 3261
ADR: 8,III

Order Now

Testitulosten erittely

Appearance Off-white to green-beige to light brown Powder or needles
Infrared spectrum Conforms
Titration with NaOH ≥60 % (As is)
Water ≤0.5 %
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR