DL-Tyrosiini 99%

Toimittaja: Thermo Fisher Scientific
Varoitus

163570250 163571000
ACRO163570250EA 82.7 EUR
ACRO163570250 ACRO163571000
DL-Tyrosiini 99%
DL-Tyrosiini
Kaava: C₉H₁₁NO₃
Kiehumispiste: 378 °C (1013 hPa)
MDL numero: MFCD00063074
CAS-numero: 556-03-6
EINECS: 209-113-1
Merck Index: 12,09970

Order Now

Testitulosten erittely

Appearance White to off-white Powder
Infrared spectrum Conforms
Titration with HClO4 98.5 to 101.5 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR