Ouabain octahydrate 96%

Toimittaja: Thermo Fisher Scientific
Vaara

161730010 161730050 161730250 161732500
ACRO161730010EA 219 EUR
ACRO161730010 ACRO161730050 ACRO161730250 ACRO161732500
Ouabain octahydrate 96%
Ouabain octahydrate

CAS No.: 11018-89-6

Kaava: C₂₉H₄₄O₁₂.8H₂O
Sulamispiste: 260 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00149240
CAS-numero: 11018-89-6
UN: 1544
ADR: 6.1,I

Order Now

Testitulosten erittely

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Loss on drying 18 % to 22 % (120°C, 2 hrs)
Solubility (1 % in water) Clear colorless
HPLC ≥95 %
Specific optical rotation -30° to -33° (20°C, 589 nm) (c=1, H2O) (on anhydrous substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR