1-Phenylcyclopentanecarboxylic acid 98%

Toimittaja: Thermo Fisher Scientific
Varoitus

161140250 161140050
ACRO161140250EA 90.7 EUR
ACRO161140250 ACRO161140050
1-Phenylcyclopentanecarboxylic acid 98%
1-Phenylcyclopentanecarboxylic acid
Kaava: C₁₂H₁₄O₂
Sulamispiste: 157…161 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00001372
CAS-numero: 77-55-4
EINECS: 201-037-7

Order Now

Testitulosten erittely

Appearance Cream to grey-brown Powder
Infrared spectrum Conforms
Melting point 157 to 161 °C
GC ≥97.5 %
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR