(4-Kloorifenyyli)etikkahappo 98%

Toimittaja: Thermo Fisher Scientific
Varoitus

160721000 160725000 160720050 160720025
ACRO160721000EA 80.5 EUR
ACRO160721000 ACRO160725000 ACRO160720050 ACRO160720025
(4-Kloorifenyyli)etikkahappo 98%
(4-Kloorifenyyli)etikkahappo
Kaava: (C₆H₄Cl)CH₂COOH
Kiehumispiste: 292 °C (1013 hPa)
Sulamispiste: 103…107 °C
Tiheys: 1,43 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00004344
CAS-numero: 1878-66-6
EINECS: 217-521-6

Order Now

Testitulosten erittely

Appearance White to cream Powder
Infrared spectrum Conforms
Melting point 103 to 107 °C
Water ≤0.2 % (K.F.)
HPLC ≥97.5 %
Appearance of solution (5% soln. in CHCl3) clear
Titration with NaOH 97.5 to 101.0 % (On dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR