D-(-)-Quinic acid 98+%

Toimittaja: Thermo Fisher Scientific
Varoitus

160500250 160501000 160505000
ACRO160500250EA 51.8 EUR
ACRO160500250 ACRO160501000 ACRO160505000
D-(-)-Quinic acid 98+%
D-(-)-Quinic acid
Kaava: C₇H₁₂O₆
Sulamispiste: 166…168 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003864
CAS-numero: 77-95-2
EINECS: 201-072-8

Order Now

Testitulosten erittely

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 162 to 168 °C
Specific optical rotation -42° to -45° (20 °C, 589 nm) (c=10, water)
Titration with NaOH ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR