1,2,3-Trimetoksibentseeni 98%

Toimittaja: Thermo Fisher Scientific

160210250 160211000
ACRO160210250EA 36.8 EUR
ACRO160210250 ACRO160211000
1,2,3-Trimetoksibentseeni 98%
1,2,3-Trimetoksibentseeni
Kaava: C₉H₁₂O₃
Kiehumispiste: 241 °C (1013 hPa)
Sulamispiste: 42…45 °C
Tiheys: 1,112 g/cm³
Leimahduspiste: 112 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00008358
CAS-numero: 634-36-6
EINECS: 211-207-2

Order Now

Testitulosten erittely

Appearance White to beige to light brown Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 42 to 47 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR