6-Nitro-m-kresoli 97%

Toimittaja: Thermo Fisher Scientific
Varoitus

160150250 160151000
ACRO160150250EA 37.1 EUR
ACRO160150250 ACRO160151000
6-Nitro-m-kresoli 97%
6-Nitro-m-kresoli
Kaava: C₇H₇NO₃
Sulamispiste: 53…56 °C
Leimahduspiste: 109 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00007111
CAS-numero: 700-38-9
EINECS: 211-843-0
UN: 2446
ADR: 6.1,III

Order Now

Testitulosten erittely

Appearance Yellow to yellow-green Crystalline powder and/or chunks
Infrared spectrum Conforms
Melting point 53 to 56 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR