Sulphur trioxide trimethylamine complex 95%

Toimittaja: Thermo Fisher Scientific
Vaara

159350250 159351000 159355000
ACRO159350250EA 40.8 EUR
ACRO159350250 ACRO159351000 ACRO159355000
Sulphur trioxide trimethylamine complex 95%
Sulphur trioxide trimethylamine complex
Kaava: C₃H₉NO₃S
Sulamispiste: 232 °C (haj. lämpötila)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00012421
CAS-numero: 3162-58-1
UN: 3261
ADR: 8,II

Order Now

Testitulosten erittely

Appearance White to off-white Powder
Infrared spectrum Conforms
Titration with NaOH ≥94 %
Iron (Fe) ≤0.001 %
Magnesium (Mg) ≤0.01 %
Potassium (K) ≤0.005 %
Heavy metals (as Pb) ≤20 ppm
Insoluble matter ≤0.02 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR