(±)-Tetrahydrofurfurylamine 97%

Toimittaja: Thermo Fisher Scientific
Varoitus

Synonyms: 2-Tetrahydrofurfurylamine

157820050 157821000 157825000
ACRO157820050EA 28.5 EUR
ACRO157820050 ACRO157821000 ACRO157825000
(±)-Tetrahydrofurfurylamine 97%
(±)-Tetrahydrofurfurylamine
Kaava: C₅H₁₁NO
Kiehumispiste: 153…154 °C (744 mmHg)
Tiheys: 0,98 g/cm³
Leimahduspiste: 45 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00005373
CAS-numero: 4795-29-3
EINECS: 225-351-9
UN: 2943
ADR: 3,III

Order Now

Testitulosten erittely

Appearance Clear colorless to dark yellow Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.4540 to 1.4570 (20°C, 589 nm)
Titration with HClO4 ≥96.0 %
UV at 400 nm A: ≤0.01
Color scale ≤10 APHA
Residue after evaporation ≤0.0005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR