Dibutyl phosphonate P 14,5-16%

Toimittaja: Thermo Fisher Scientific
Varoitus

155061000 155065000
ACRO155061000EA 36.8 EUR
ACRO155061000 ACRO155065000
Dibutyl phosphonate P 14,5-16%
Dibutyl phosphonate
Kaava: C₈H₁₉O₃P
Kiehumispiste: 118…119 °C (11 mmHg)
Sulamispiste: –57 °C
Tiheys: 0,995 g/cm³
Leimahduspiste: 121 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00066633
CAS-numero: 1809-19-4
EINECS: 217-316-1

Order Now

Testitulosten erittely

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4225 to 1.4255 (20°C, 589 nm)
Assay 14.5 to 16 % (P)
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR