tert-Butyl bromoacetate 99%

Toimittaja: Thermo Fisher Scientific
Varoitus

154340100 154340500 154342500
ACRO154340100EA 48.8 EUR
ACRO154340100 ACRO154340500 ACRO154342500
tert-Butyl bromoacetate 99%
tert-Butyl bromoacetate
Kaava: BrCH₂COOC(CH₃)₂
Kiehumispiste: 50 °C (10 mmHg)
Tiheys: 1,338 g/cm³
Leimahduspiste: 49 °C
MDL numero: MFCD00000188
CAS-numero: 5292-43-3
EINECS: 226-133-6
UN: 1993

Order Now

Testitulosten erittely

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4440 to 1.4460 (20°C, 589 nm)
GC ≥98.5 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR