Luminol (3-Aminophthalhydrazide) 98%, pure

Toimittaja: Thermo Fisher Scientific
Varoitus

Synonyms: NSC 5064

153850050 153850250 153850010 153851000
ACRO153850050EA 105 EUR
ACRO153850050 ACRO153850250 ACRO153850010 ACRO153851000
Luminol (3-Aminophthalhydrazide) 98%, pure
Luminol (3-Aminophthalhydrazide)
Kaava: C₈H₇N₃O₂
Sulamispiste: 319…320 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00006890
CAS-numero: 521-31-3
EINECS: 208-309-4

Order Now

Testitulosten erittely

Appearance Yellow to yellow-green to beige Powder and/or chunks
Infrared spectrum Conforms
Solubility (2 % in 1 N NaOH) Clear yellow to yellow-green
Titration excess NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR