Cysteaminium chloride 98%

Toimittaja: Thermo Fisher Scientific
Varoitus

153770250 153771000 153770050 153775000
ACRO153770250EA 66.5 EUR
ACRO153770250 ACRO153771000 ACRO153770050 ACRO153775000
Cysteaminium chloride 98%
Cysteaminium chloride
Kaava: C₂H₇NS·HCl
Sulamispiste: 66…70 °C
Storage Temperature: Jääkaappi
MDL numero: MFCD00012904
CAS-numero: 156-57-0
EINECS: 205-858-1
UN: 3335
ADR: 9,III

Order Now

Testitulosten erittely

Appearance White Crystalline powder or crystals and/or lumps
Infrared spectrum Conforms
Melting point 66 to 70 °C
Loss on drying ≤1 % (50 °C, 4 h, 100 mmHg)
Titration with KIO3 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR