Neokuproiinihydrokloridimonohydraatii 99%

Toimittaja: Thermo Fisher Scientific
Varoitus

153310050 153310250 153311000
ACRO153310050EA 102 EUR
ACRO153310050 ACRO153310250 ACRO153311000
Neokuproiinihydrokloridimonohydraatii 99%
Neokuproiinihydrokloridimonohydraatii
Kaava: C₁₄H₁₃ClN₂·1H₂O
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00012645
CAS-numero: 303136-82-5
EINECS: 230-732-8

Order Now

Testitulosten erittely

Appearance Light yellow Crystalline powder
Infrared spectrum Conforms
Sulfated ash ≤0.1 %
Titration with HClO4 ≥98.5 %
Suitability (for Cu determination) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR