2,2'-dihydroksibifenyyli 99%

Toimittaja: Thermo Fisher Scientific
Varoitus

151991000 151995000
ACRO151991000EA 56.2 EUR
ACRO151991000 ACRO151995000
2,2'-dihydroksibifenyyli 99%
2,2'-dihydroksibifenyyli
Kaava: C₁₂H₁₀O₂
Kiehumispiste: 315 °C (1013 hPa)
Sulamispiste: 108…111 °C
Tiheys: 0,83 g/cm³
Leimahduspiste: 154 °C
MDL numero: MFCD00002210
CAS-numero: 1806-29-7
EINECS: 217-303-0

Order Now

Testitulosten erittely

Appearance Beige to light grey Powder
Infrared spectrum Conforms
Melting point 108 to 111 °C
Loss on drying ≤0.5 %
GC ≥98.5 %
Iron (Fe) ≤15 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR