1,6-Heptadien-4-ol 97%

Toimittaja: Thermo Fisher Scientific
Varoitus

150090050 150090250
ACRO150090050EA 109 EUR
ACRO150090050 ACRO150090250
1,6-Heptadien-4-ol 97%
1,6-Heptadien-4-ol
Kaava: C₇H₁₂O
Kiehumispiste: 151 °C
Tiheys: 0,86 g/cm³
Leimahduspiste: 40 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00008663
CAS-numero: 2883-45-6
EINECS: 220-742-0
UN: 1987
ADR: 3,III

Order Now

Testitulosten erittely

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4490 to 1.4530 (20°C, 589 nm)
GC ≥96.0 %
Residue after ignition 35 % to 50 %
pH 5 to 8 (1 % solution)
Specific optical rotation +75° to +105° (20°C, 589 nm) (6% in water)
Specific viscosity 0.018 to 0.032 (1.0M in NaCl, 25°C)
Sulfur (S) 16 to 20 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR