2-Metoksietanoli 99.5+% for analysis

Toimittaja: Thermo Fisher Scientific
Vaara

149360010 149360025
ACRO149360010EA 93.6 EUR
ACRO149360010 ACRO149360025
2-Metoksietanoli 99.5+% for analysis
2-Metoksietanoli
Kaava: C₃H₈O₂
Kiehumispiste: 124 °C (760 mmHg)
Sulamispiste: –85 °C
Tiheys: 0,96 g/cm³
Leimahduspiste: 38 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00002867
CAS-numero: 109-86-4
EINECS: 203-713-7
UN: 1188
ADR: 3,III
Merck Index: 12,06118

Order Now

Testitulosten erittely

Appearance Clear liquid
Water ≤0.1 % (Coulometric)
GC ≥99.5 %
Color scale ≤10 APHA
Non-volatile matter ≤0.002 %
Acidity ≤0.001 meq/g
Alkalinity ≤0.0005 meq/g
Substance colored by H2SO4 Passes test
Aluminium (Al) ≤5 ppm
Barium (Ba) ≤1 ppm
Cadmium (Cd) ≤0.05 ppm
Calcium (Ca) ≤0.5 ppm
Chromium (Cr) ≤0.02 ppm
Copper (Cu) ≤0.02 ppm
Iron (Fe) ≤0.5 ppm
Lead (Pb) ≤0.1 ppm
Magnesium (Mg) ≤0.1 ppm
Manganese (Mn) ≤0.02 ppm
Nickel (Ni) ≤0.02 ppm
Zinc (Zn) ≤0.1 ppm
Aldehyde (C2H4O) ≤0.003 %
Peroxides ≤20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR