Aminoguanidinium hydrogen carbonate 98.5%

Toimittaja: Thermo Fisher Scientific
Vaara

149060010 149062500 149060051
ACRO149060010EA 88.6 EUR
ACRO149060010 ACRO149062500 ACRO149060051
Aminoguanidinium hydrogen carbonate 98.5%
Aminoguanidinium hydrogen carbonate
Kaava: C₂H₈N₄O₃
Sulamispiste: 125 °C (haj. lämpötila)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00012949
CAS-numero: 2582-30-1
EINECS: 219-956-7
UN: 3077
ADR: 9,III

Order Now

Testitulosten erittely

Appearance White to cream to light yellow Crystalline powder
Infrared spectrum Conforms
Residue after ignition ≤0.3 %
Titration with HClO4 98.0 to 102.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR