1,1,1,3,3,3-Heksafluori-2-propanoli 99.5+%, pure

Toimittaja: Thermo Fisher Scientific
Vaara

147540050 147540250 147541000 147540025 147545000
ACRO147540050EA 46.3 EUR
ACRO147540050 ACRO147540250 ACRO147541000 ACRO147540025 ACRO147545000
1,1,1,3,3,3-Heksafluori-2-propanoli 99.5+%, pure
1,1,1,3,3,3-Heksafluori-2-propanoli
Kaava: C₃H₂F₆O
Kiehumispiste: 59 °C (760 mmHg)
Sulamispiste: –4 °C
Tiheys: 1,596 g/cm³
Leimahduspiste: >100 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011651
CAS-numero: 920-66-1
EINECS: 213-059-4
UN: 3265
ADR: 8,I
Merck Index: 13,09598

Order Now

Testitulosten erittely

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.3 %
GC ≥99.50 %
Specific optical rotation +170° to +180° (20°C, 589 nm) (C=2 in CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR