3,4,5-Trimetoksianiliini 97%

Toimittaja: Thermo Fisher Scientific
Varoitus

139900100 139900250
ACRO139900100EA 61.1 EUR
ACRO139900100 ACRO139900250
3,4,5-Trimetoksianiliini 97%
3,4,5-Trimetoksianiliini
Kaava: C₉H₁₃NO₃
Kiehumispiste: 293 °C (1013 hPa)
Sulamispiste: 110…113 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00008393
CAS-numero: 24313-88-0
EINECS: 246-154-4
UN: 2811
ADR: 6.1,III

Order Now

Testitulosten erittely

Appearance Off-white to beige to light brown Powder
Infrared spectrum Conforms
Melting point 108 to 113 °C
Titration with HClO4 ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR