2-Thenoic acid 99%

Toimittaja: Thermo Fisher Scientific
Varoitus

138841000 138840050 138840250 138845000
ACRO138841000EA 68 EUR
ACRO138841000 ACRO138840050 ACRO138840250 ACRO138845000
2-Thenoic acid 99%
2-Thenoic acid
Kaava: C₅H₄O₂S
Kiehumispiste: 260 °C (760 mmHg)
Sulamispiste: 126…130 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00005437
CAS-numero: 527-72-0
EINECS: 208-423-4
Merck Index: 12,09491

Order Now

Testitulosten erittely

Appearance White to cream to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 126 to 130 °C
GC ≥98.5 %
Titration with NaOH 98.5 to 101.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR