4-Formylmorpholine 99+%

Toimittaja: Thermo Fisher Scientific

Synonyms: Morpholine-4-carbaldehyde, N-Formylmorpholine

133800050 133801000 133805000
ACRO133800050EA 25.7 EUR
ACRO133800050 ACRO133801000 ACRO133805000
4-Formylmorpholine 99+%
4-Formylmorpholine
Kaava: C₅H₉NO₂
Sulamispiste: 23 °C
Tiheys: 1,14 g/cm³
Leimahduspiste: 113 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00006170
CAS-numero: 4394-85-8
EINECS: 224-518-3

Order Now

Testitulosten erittely

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4855 to 1.4875 (20°C, 589 nm)
GC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR