Semikarbatsidi-HCl 99+%

Toimittaja: Thermo Fisher Scientific
Vaara

132641000 132645000 132640025
ACRO132641000EA 41.6 EUR
ACRO132641000 ACRO132645000 ACRO132640025
Semikarbatsidi-HCl 99+%
Semikarbatsidi-HCl
Kaava: CH₆ClN₃O
Sulamispiste: 170…178 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00013009
CAS-numero: 563-41-7
EINECS: 209-247-0
UN: 2811
ADR: 6.1,III
Merck Index: 12,08588

Order Now

Testitulosten erittely

Appearance White Crystalline powder and/or chunks
Infrared spectrum Conforms
Melting point 170 to 178 °C
Sulfated ash ≤0.1 %
Solubility (10 % in water) Clear colorless
Titration with KIO3 ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR