2-Fenyylipropaanihappo 98%

Toimittaja: Thermo Fisher Scientific
Varoitus

130870100 130870250 130871000
ACRO130870100EA 66.3 EUR
ACRO130870100 ACRO130870250 ACRO130871000
2-Fenyylipropaanihappo 98%
2-Fenyylipropaanihappo
Kaava: C₉H₁₀O₂
Kiehumispiste: 260…262 °C (760 mmHg)
Sulamispiste: 5 °C
Tiheys: 1,1 g/cm³
Leimahduspiste: 148 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00002650
CAS-numero: 492-37-5
EINECS: 207-752-0

Order Now

Testitulosten erittely

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5210 to 1.5250 (20°C, 589 nm)
GC ≥97.5 %
Titration with NaOH 97.5 to 102.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR