2-Phenylindole 99%

Toimittaja: Thermo Fisher Scientific

130680050 130681000
ACRO130680050EA 31.8 EUR
ACRO130680050 ACRO130681000
2-Phenylindole 99%
2-Phenylindole
Kaava: C₁₄H₁₁N
Kiehumispiste: 250 °C (10 mmHg)
Sulamispiste: 188…191 °C
MDL numero: MFCD00005608
CAS-numero: 948-65-2
EINECS: 213-436-3

Order Now

Testitulosten erittely

Appearance Off-white to beige to light green Powder
Infrared spectrum Conforms
Melting point 188 to 191 °C
Water ≤0.5 % (K.F.)
Sulfated ash ≤0.2 %
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR