4-Phenoxyaniline 97%

Toimittaja: Thermo Fisher Scientific
Varoitus

Synonyms: 1-Amino-4-phenoxybenzol

130231000 130230050 130235000
ACRO130231000EA 74.3 EUR
ACRO130231000 ACRO130230050 ACRO130235000
4-Phenoxyaniline 97%
4-Phenoxyaniline
Kaava: C₁₂H₁₁NO
Kiehumispiste: 140 °C (3hPa)
Sulamispiste: 82…86 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00007862
CAS-numero: 139-59-3
EINECS: 205-367-2
UN: 3077
ADR: 9,III

Order Now

Testitulosten erittely

Appearance Brown to green-brown Powder or flakes
Infrared spectrum Conforms
Melting point 82 to 86 °C
Titration with HClO4 ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR